Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTRH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554353
Description
Immunogen sequence: VYLVHDELDK PLGRLALKLG GSARGHNGVR SCISCLNSNA MPRLRVGIGR PAHPEAVQAH VLGCFSPAEQ ELLPLLLDRA TD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
PTRH1 (Peptidyl-TRNA Hydrolase 1 Homolog) is a Protein Coding gene. Diseases associated with PTRH1 include Vascular Skin Disease and Early Infantile Epileptic Encephalopathy. Gene Ontology (GO) annotations related to this gene include aminoacyl-tRNA hydrolase activity.
Specifications
PTRH1 | |
Polyclonal | |
Unconjugated | |
PTRH1 | |
2210013M04Rik; AW046744; C9orf115; peptidyl-tRNA hydrolase 1 homolog; peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae); probable peptidyl-tRNA hydrolase; PTH; PTH1; Ptrh1; RGD1310165 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
138428 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86Y79 | |
PTRH1 | |
Recombinant protein corresponding to Human PTRH1. Recombinant protein control fragment (Product #RP-93080). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction