Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTRH2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055963AF405
Description
PTRH2 Polyclonal specifically detects PTRH2 in Human, Mouse samples. It is validated for Western Blot.Specifications
PTRH2 | |
Polyclonal | |
Western Blot | |
Bcl-2 inhibitor of transcription 1, BIT1CGI-147, EC 3.1.1.29, mitochondrial, peptidyl-tRNA hydrolase 2, PTH2FLJ32471 | |
Rabbit | |
Affinity Purified | |
RUO | |
Bit1 (IN) | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PTRH2 | |
Antibody was raised against a 16 amino acid synthetic peptide from near the center of human PTRH2. The immunogen is located within amino acids 80 - 130 of PTRH2. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction