Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTS Antibody (02), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306548F
Description
PTS Monoclonal antibody specifically detects PTS in Human samples. It is validated for ELISASpecifications
PTS | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
02 | |
ELISA | |
6-pyruvoyl tetrahydrobiopterin synthase, 6-pyruvoyltetrahydropterin synthase, EC 4.2.3.12, FLJ97081, PTPSPTP synthase | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PTS (Q03393; Met1-Glu145). | |
0.1 mL | |
Phospho Specific | |
5805 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction