Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTS, p Ser19 Antibody (SR1297), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321604100UL
Description
PTS Monoclonal antibody specifically detects PTS in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
PTS | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1297 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
6-pyruvoyl tetrahydrobiopterin synthase, 6-pyruvoyltetrahydropterin synthase, EC 4.2.3.12, FLJ97081, PTPSPTP synthase | |
A synthesized peptide derived from human phospho-PTS [Ser19] (Uniprot #: Q03393) | |
100 μg | |
Phospho Specific | |
5805 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction