Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTTG1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP269985
Description
PTTG1 Polyclonal specifically detects PTTG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
PTTG1 | |
Polyclonal | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Peptide ELISA 1:100-1:2000 | |
EAP1MGC126883, Esp1-associated protein, ESP1-associated protein 1, hPTTG, pituitary tumor-transforming 1, Pituitary tumor-transforming gene 1 protein, PTTGMGC138276, securin, Tumor-transforming protein 1, TUTR1HPTTG | |
Goat | |
Antigen Affinity-purified | |
Breast Cancer, Cell Cycle and Replication, Mitotic Regulators | |
9232 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
TBS (pH 7.3) with 0.5% BSA with 0.02% Sodium Azide | |
PTTG1 | |
Peptide with sequence C-ILDEERELEKLFQ, from the internal region of the protein sequence according to NP_004210.1. | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction