Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ PTV Liner Seal Pre-Seat Tool
SDP

Catalog No. 06611189 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-611-189 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-189 Supplier Restek™ Supplier No. 22352
May include imposed supplier surcharges.
Add to Cart
Add to Cart

For Gerstel CIS 3 and CIS 4 PTV Inlets

Specifications

Type PTV Liner Seal Pre-Seat Tool
Color Silver
For Use With (Application) Gerstel CIS 3 and CIS 4 PTV Inlets
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.