Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTX3 Monoclonal Antibody (03), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | PTX3 |
---|---|
Clone | 03 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543537
|
Invitrogen™
MA543537 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543031
|
Invitrogen™
MA543031 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human PTX3/Pentraxin 3/TSG-14
Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of "e;long pentraxins"e;, which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.Specifications
PTX3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14 | |
PTX3 | |
IgG1 | |
Protein A | |
Antibody |
03 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P26022 | |
5806 | |
Recombinant Human PTX3/Pentraxin 3/TSG-14 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
PTX3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title