Learn More
Invitrogen™ PTX3 Monoclonal Antibody (2C3)

Description
Product is supplied at 1 mL (100 μL/mL). Application Notes: Western blot- A non-reduced sample treatment and SDS-Page was used. The band size is 400 kDa. Flow cytometry - This antibody stains the intra and extracellular domain of Pentraxin 3. For intracellular staining cells were permeabilized with buffer containing triton. Transfected cells were used as a positive control.
Specifications
Specifications
| Antigen | PTX3 |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2C3 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene | PTX3 |
| Gene Accession No. | P48759 |
| Gene Alias | AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.