Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTX3 Monoclonal Antibody (MNB4)
GREENER_CHOICE

Catalog No. FERMA549098
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549098 Supplier Invitrogen™ Supplier No. MA549098
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PTX3 Monoclonal Antibody for Flow, ICC/IF, IHC (F), IHC (P), Inhibition, IP, Western Blot

Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of 'long pentraxins', which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTX3
Applications Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MNB4
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene PTX3
Gene Accession No. P26022
Gene Alias AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14
Gene Symbols PTX3
Host Species Rat
Immunogen Human recombinant PTX3.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5806
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.