Learn More
Invitrogen™ PTX3 Polyclonal Antibody

Description
Immunogen sequence: GFDETLAFSG RLTGFNIWDS VLSNEEIRET GGAESCHIRG NIVGWGVTEI QPHGGAQYVS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 90%.
Specifications
Specifications
| Antigen | PTX3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PTX3 |
| Gene Accession No. | P26022 |
| Gene Alias | AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14 |
| Gene Symbols | PTX3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.