Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545841
Description
Peptide sequence: LGERSQQRAR ERKAHKAQRD ALQDSLARLE GLVHSQGARL AALEGRLPVA.
PTX4 belongs to the pentraxin (PTX) superfamily of multifunctional conserved proteins. Pentraxins are characterized by a cyclic multimeric structure and a conserved C-terminal pentraxin domain of approximately 200 amino acids. All pentraxins have a conserved 8-amino acid sequence, HxCxS/TWxS, in which x is any amino acid, within their pentraxin domain. Pentraxins are divided into short proteins, such as C-reactive protein and serum amyloid P protein, and long proteins, such as PTX3 and PTX4. Some PTXs are part of the humoral arm of innate immunity and behave as functional ancestors of antibodies by mediating agglutination, complement activation, and opsonization.
Specifications
PTX4 | |
Polyclonal | |
Unconjugated | |
PTX4 | |
C16orf38; long pentraxin 4; neuronal pentraxin-like protein C16orf38; pentraxin 4; pentraxin 4, long; pentraxin-4; PTX4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
390667 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96A99 | |
PTX4 | |
Synthetic peptide directed towards the N-terminal of human PTX4 (aa 129-178). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction