Learn More
Invitrogen™ PU.1 Monoclonal Antibody (5C7)

Description
MA5-17178 targets SPI1 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17178 immunogen is purified recombinant fragment of human SPI1 (amino acids: 124-271) expressed in E. Coli. MA5-17178 detects SPI1 which has a predicted molecular weight of approximately 42kDa.
Specifications
Specifications
| Antigen | PU.1 |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 5C7 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | SPI1 |
| Gene Accession No. | P17947 |
| Gene Alias | 31 kDa transforming protein; 31 kDa-transforming protein; 31kDa Transforming Protein; Dis1; Dis-1; hCG_25181; hematopoietic transcription factor PU.1; OF; Pu.1; SFFV proviral integration 1; SFFV proviral integration 1 protein; Sfpi1; Sfpi-1; Spi1; SPI-1; Spi-1 proto-oncogene; SPIA; SPI-A; spleen focus forming virus (SFFV) proviral integration oncogene; spleen focus forming virus (SFFV) proviral integration oncogene spi1; spleen focus forming virus proviral integration oncogene spi1; Tcfpu1; Tfpu.1; Transcription factor PU.1; Transcription Factor spi1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.