Learn More
Puck Set
Click to view available options
:
Balloon pucks, 7 and 9cm
Description
Includes
Two small pucks, one large puck (twice the mass of the small pucks), six balloons, three stoppers
- Study collisions in two dimensions and demonstrate both uniform velocity and accelerated motion
- The pucks ride on a film of air produced by their integral inflated balloons
Specifications
Specifications
| Product Type | Balloon Pucks |
| For Use With (Application) | Demonstrate collisions in two dimensions |
| Includes | Two small pucks, one large puck (twice the mass of the small pucks), six balloons, three stoppers |