Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUF60 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PUF60 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
PUF60 Polyclonal specifically detects PUF60 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PUF60 | |
Polyclonal | |
Purified | |
RUO | |
FBP interacting repressor, FBP-interacting repressor, FIRpoly-U binding splicing factor PUF60, FLJ31379, FUSE-binding protein-interacting repressor, poly(U)-binding-splicing factor PUF60, poly-U binding splicing factor 60KDa, Ro ribonucleoprotein binding protein 1, Ro ribonucleoprotein-binding protein 1, Ro-binding protein 1, roBP1, ROBPI, siah binding protein 1,60 kDa poly(U)-binding-splicing factor, Siah-binding protein 1, siah-BP1, SIAHBP1pyrimidine tract binding splicing factor | |
PUF60 | |
IgG | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Q9UHX1-2 | |
22827 | |
Synthetic peptides corresponding to PUF60(poly-U binding splicing factor 60KDa) The peptide sequence was selected from the C terminal of PUF60. Peptide sequence PPIPVTIPSVGVVNPILASPPTLGLLEPKKEKEEEELFPESERPEMLSEQ. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title