Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PUF60 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584430 Supplier Invitrogen™ Supplier No. PA584430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PIIDQLAEEA RAFNRIYVAS VHQDLSDDDI KSVFEAFGKI KSCTLARDPT TGKHKGYGFI EYEKAQSSQD AVSSMNLFDL GG Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

PUF60 encodes a protein that is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUF60
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Puf60
Gene Accession No. Q9UHX1
Gene Alias 2410104I19Rik; 2810454F19Rik; 60 kDa poly(U)-binding-splicing factor; FBP interacting repressor; FBP-interacting repressor; FIR; FLJ31379; FUSE-binding protein-interacting repressor; LOW QUALITY PROTEIN: poly(U)-binding-splicing factor PUF60; poly(U) binding splicing factor 60; poly(U) binding splicing factor 60KDa; poly(U)-binding-splicing factor PUF60; poly-U binding splicing factor 60; poly-U binding splicing factor 60K; poly-U binding splicing factor 60KDa; PUF60; pyrimidine tract binding splicing factor; RNA-binding protein Siah-BP; Ro ribonucleoprotein-binding protein 1; Ro-binding protein 1; RoBP1; ROBPI; Siah binding protein 1; siah binding protein 1; FBP interacting repressor; pyrimidine tract binding splicing factor; Ro ribonucleoprotein-binding protein 1; siah-binding protein 1; SIAHBP1; Siah-BP1; similar to fuse-binding protein-interacting repressor isoform a; VRJS
Gene Symbols Puf60
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PUF60. Recombinant protein control fragment (Product #RP-104072).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.