Learn More
PUF60 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302818A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
The protein encoded by this gene is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene.Specifications
PUF60 | |
Polyclonal | |
Unconjugated | |
PUF60 | |
PUF60; SIAHBP1; FIR; fuse-binding protein-interacting repressor; RoBPI; siah binding protein 1; poly-U binding splicing factor PUF60; Ro ribonucleoprotein-binding protein 1; pyrimidine tract binding splicing factor; poly-U binding splicing factor 60KDa; FLJ31379; FBP interacting repressor; Ro ribonucleoprotein binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22827, 67959 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q3UEB3, Q9UHX1 | |
PUF60 | |
Between 400 and 450 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.