Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uniform Technology Pullover Eyes Only Hood Tricot, White

Catalog No. 17779023 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-779-023 Small
17-779-022 X-Small
17-779-024 Medium
17-779-025 Large
17-779-026 2X-Large
17-779-027 3X-Large
17-779-033 X-Large
17-779-036 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-779-023 Supplier Uniform Technology Supplier No. CH283209WHS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Uniform Technology Pullover Eyes Only Hood with shoulder drape and built-in 2 layer mask in 2-ply tricot.

  • Hood
  • Pull-over eyes only
  • Shoulder drape
  • Built-in 2 layer mask in 2-ply tricot
  • Vertical 2-ply #35 white ear vents
  • Eyeglass slits
  • Vertical and horizontal snaps in the back
  • Ideal for General CE Spaces/Applications (Non-ESD sensitive Medical Device, Electronics & Aerospace, Food Processing/Handling or Packaging, Cultivation Industry, University Labs, Imaging/Film coating Manufactures, General Medical/Dental Facilities

Specifications

Product Type Cleanroom Hoods
Size Small
Material Polyester, Carbon
Color White
Closure Type Pull-over
Disposable Reusable
Latex Free Yes
Seam Type Bound
Sterility Non-sterile
Length (Metric) 45.72 cm
Length (Metric) Facepiece 45.72 cm
For Use With (Application) General Cleanroom Spaces
Resistance Anti-static
Cleanroom Class Class 10 (ISO 4)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.