Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUM1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100259AF488
Description
PUM1 Polyclonal antibody specifically detects PUM1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PUM1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
HSPUM, KIAA0099PUML1, PUMH, PUMH1pumilio homolog 1, pumilio (Drosophila) homolog 1, pumilio homolog 1 (Drosophila), Pumilio-1 | |
The immunogen recognized by this antibody maps to a region between residues 225 and 275 of human Pumilio 1 using the numbering given in entry NP_055491.1 (GeneID 9698). | |
0.1 mL | |
Cell Biology, Stem Cells | |
9698 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction