Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PUM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589209 Supplier Invitrogen™ Supplier No. PA589209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSVACVLKRK AVLWQDSFSP HLKHHPQEPA NPNMPVVLTS GTGSQAQPQP AANQALAAGT HSSPVPGSIG VAGRSQDDAM VDYFFQRQHG EQLGGGGSGG GGYNNSKHRW PTGDNIHAEH QVRSMDELNH; Positive Samples: HepG2, SH-SY5Y, Mouse liver; Cellular Location: Cytoplasm, Cytoplasmic granule, P-body.

PUM1 (pumilio homolog 1) is a sequence-specific RNA-binding protein that acts as a post-transcriptional repressor by binding the 3'-UTR of mRNA targets. It is a member of the PUF family comprised of evolutionarily conserved RNA-binding proteins. PUM1 binds to an RNA consensus sequence, the Pumilio Response Element (PRE), and 5'-UGUANAUA-3' that is related to the Nanos Response Element (NRE). PUM1 mediates post-transcriptonal repression of transcripts via different mechanisms - acts via direct recruitment of the CCR4-POP2-NOT deadenylase leading to translational inhibition and mRNA degradation. Mutations to the gene can result in spinocerebellar ataxia 47.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUM1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PUM1
Gene Accession No. Q14671, Q80U78
Gene Alias AA517475; HsPUM; KIAA0099; mKIAA0099; PUM1; PUMH; PUMH1; pumilio 1; pumilio homolog 1; pumilio RNA binding family member 1; pumilio RNA-binding family member 1; pumilio-1; PUML1; Pumm
Gene Symbols PUM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human Pumilio 1 (NP_0554911).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80912, 9698
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.