Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUM2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298727MFV610
Description
PUM2 Polyclonal antibody specifically detects PUM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
PUM2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
FLJ36528, KIAA0235PUMH2PUML2, MGC138251, MGC138253, pumilio (Drosphila) homolog 2, pumilio homolog 2, pumilio homolog 2 (Drosophila), Pumilio-2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PUM2. | |
0.1 mL | |
Stem Cell Markers, Translation Control | |
23369 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction