Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB100387
Description
PUM2 Polyclonal specifically detects PUM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PUM2 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
FLJ36528, KIAA0235PUMH2PUML2, MGC138251, MGC138253, pumilio (Drosphila) homolog 2, pumilio homolog 2, pumilio homolog 2 (Drosophila), Pumilio-2 | |
Rabbit | |
114 kDa | |
100 μL | |
Stem Cell Markers, Translation Control | |
23369 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:10000-1:20000, Immunoprecipitation 5-10 ug/mg lysate | |
Q8TB72 | |
PUM2 | |
The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Pumilio 2 using the numbering given in entry NP_056132.1 (GeneID 23369). | |
Affinity Purified | |
RUO | |
Primary | |
Rat reactivity reported in scientific literature (PMID: 25097251). Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eatingmacaque, African elephant, Small-eared galago and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction