Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUM2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297161PCP
Description
PUM2 Polyclonal antibody specifically detects PUM2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
PUM2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
FLJ36528, KIAA0235PUMH2PUML2, MGC138251, MGC138253, pumilio (Drosphila) homolog 2, pumilio homolog 2, pumilio homolog 2 (Drosophila), Pumilio-2 | |
Produced in rabbits immunized with E. coli-derived Human PUM2 fragment. | |
0.1 mL | |
Stem Cell Markers, Translation Control | |
23369 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction