Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PUM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518355
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518355 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518355 Supplier Invitrogen™ Supplier No. PA518355
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PUM2
Gene Accession No. Q8TB72
Gene Alias 5730503J23Rik; FLJ36528; KIAA0235; MGC138251; MGC138253; mKIAA0235; OTTHUMP00000200539; PUM1; Pum2; PUMH2; pumilio 2; pumilio homolog 2; pumilio RNA binding family member 2; pumilio RNA-binding family member 2; pumilio-2; PUML2; Pumm2
Gene Symbols PUM2
Host Species Goat
Immunogen Synthetic peptide sequence (DQNRDLKQGDDDDSK) corresponding to the internal amino acids of PUM2 (aa 141-156).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23369
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.