Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PUM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556436
Description
Immunogen sequence: ASPTEVVERL GPNTNPSEGL GPLPNPTANK PLVEEFSNPE TQNLDAMEQV GLESLQF Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
Specifications
| PUM2 | |
| Polyclonal | |
| Unconjugated | |
| PUM2 | |
| 5730503J23Rik; FLJ36528; KIAA0235; MGC138251; MGC138253; mKIAA0235; OTTHUMP00000200539; PUM1; Pum2; PUMH2; pumilio 2; pumilio homolog 2; pumilio RNA binding family member 2; pumilio RNA-binding family member 2; pumilio-2; PUML2; Pumm2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23369 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TB72 | |
| PUM2 | |
| Recombinant protein corresponding to Human PUM2. Recombinant protein control fragment (Product #RP-95963). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction