Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUM2 Rabbit anti-Human, Mouse, Rat, Clone: 7M1B1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PUM2 |
---|---|
Clone | 7M1B1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PUM2 Monoclonal antibody specifically detects PUM2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PUM2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Stem Cell Markers, Translation Control | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
23369 | |
IgG | |
Affinity purified |
7M1B1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ36528, KIAA0235PUMH2PUML2, MGC138251, MGC138253, pumilio (Drosphila) homolog 2, pumilio homolog 2, pumilio homolog 2 (Drosophila), Pumilio-2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PUM2 (Q8TB72). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title