Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUM3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP217042
Description
PUM3 Polyclonal specifically detects PUM3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KIAA0020 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
HBV X-transactivated gene 5 protein, HLA-HA8MGC8749, KIAA0020, KIAA0020 protein, Minor histocompatibility antigen HA-8, PEN, penguin homolog, protein 5 transactivated by hepatitis B virus X antigen (HBxAg), PUF6, XTP5HBV XAg-transactivated protein 5 | |
Rabbit | |
74 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Bovine (89%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q15397 | |
KIAA0020 | |
Recombinant protein encompassing a sequence within the C-terminus region of human KIAA0020. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
9933 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction