Learn More
Invitrogen™ PUMA alpha Polyclonal Antibody

Description
Predicted to react with mouse based on sequence homology. Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for this product is human breast carcinoma. In Western blot, this antibody detects a band at ~23kDa in K562 cell lysate. Rabbit anti PUMA antibody recognizes the p53 - upregulated modulator of apoptosis, also known as Bcl-2 binding component 3, JFY-1 or Bcl-2.
Specifications
Specifications
| Antigen | PUMA alpha |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BBC3 |
| Gene Accession No. | Q9BXH1 |
| Gene Alias | BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; FLJ42994; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1 |
| Gene Symbols | BBC3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.