Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUMA Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 6 publications

$170.97 - $418.55

Specifications

Antigen PUMA
Concentration 1.0 mg/ml
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:250, Immunohistochemistry-Paraffin reported in scientific literature (Karst et al (2005))
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056370T
SDP
View Documents
Novus Biologicals
NB10056370SS
0.025 mg
Each for $170.97
Only null left
Add to Cart
 
NB10056370
SDP
View Documents
Novus Biologicals
NB10056370
0.1 mg
Each for $418.55
Only null left
Add to Cart
 
Description

Description

PUMA Polyclonal specifically detects PUMA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PUMA
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:250, Immunohistochemistry-Paraffin reported in scientific literature (Karst et al (2005))
Polyclonal
Purified
RUO
Human, Mouse
BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994
BBC3
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human (PUMA a and b). Human PUMA a is a 193 amino acid protein with a predicted molecular weight of approximately 22 kD. Human PUMA b is a 131 amino acid protein with a predicted molecular weight of approximately 15 kD.
1.0 mg/ml
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Cancer, DNA Double Strand Break Repair, Signal Transduction, Tumor Suppressors
PBS with 0.05% Sodium Azide
27113
A portion of amino acids 186-199 (PLPRGHRAPEMEPN) of human PUMA a was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.