Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUMA Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500261AF532
Description
PUMA Polyclonal specifically detects PUMA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PUMA | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994 | |
Rabbit | |
Affinity Purified | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
27113.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
BBC3 | |
A synthetic peptide made to a region within the C-terminus of the human PUMA protein sequence (between residues 150-193). [UniProtNo. Q9BXH1] | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction