Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUMA Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500261AF594
Description
PUMA Polyclonal specifically detects PUMA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PUMA | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994 | |
| Rabbit | |
| Affinity Purified | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 27113.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BBC3 | |
| A synthetic peptide made to a region within the C-terminus of the human PUMA protein sequence (between residues 150-193). [UniProtNo. Q9BXH1] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction