Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUMA Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500230F
Description
PUMA Polyclonal antibody specifically detects PUMA in Human samples. It is validated for Western BlotSpecifications
PUMA | |
Polyclonal | |
Western Blot | |
BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994 | |
Reacts with a 17 residue sequence [EQHLESPVPSAPGALAG] found in the exon-3-encoded region in the human PUMA-alpha and PUMA-beta forms of the protein. Recognizes both forms of the protein. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
27113 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction