Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUMA Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500261F
Description
PUMA Polyclonal specifically detects PUMA in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PUMA | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
Q9BXH1 | |
BBC3 | |
A synthetic peptide made to a region within the C-terminus of the human PUMA protein sequence (between residues 150-193). [UniProt# Q9BXH1] | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
FITC | |
PBS with 0.05% Sodium Azide | |
BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994 | |
Rabbit | |
16 kDa | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
27113 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction