Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUMA Antibody, Novus Biologicals™
SDP

Catalog No. p-5082120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056623T 0.025 mg
NB10056623 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056623T Supplier Novus Biologicals Supplier No. NB10056623SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PUMA Polyclonal specifically detects PUMA in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen PUMA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_001120712.1
Gene Alias BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994
Gene Symbols BBC3
Host Species Rabbit
Immunogen A synthetic peptide (AAPPLTLEGPVQSHHGTP) corresponding to amino acids 36-53 of human PUMA d (GenBank no. gb AAK39544.1 AF354656_1 ) and amino acids 196-213 of human PUMA g (GenBank no ref NP_001120712.1 ) was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27113
Test Specificity The amino acid sequence used as immunogen has 100% identity in PUMAd and PUMAg, and 59% identity in PUMAa. For additional information on PUMA isoforms and their sequences, see Nakano et al. 2001.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.