Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUMA Antibody (SR1298), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321667100UL
Description
PUMA Monoclonal antibody specifically detects PUMA in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
PUMA | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1298 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994 | |
A synthesized peptide derived from human PUMA (Uniprot #: Q9BXH1) | |
100 μg | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
27113 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction