Learn More
Invitrogen™ PUMA delta/gamma Polyclonal Antibody

Description
Some cross reactivity is seen with PUMA alpha. This sequence is 59% homologus to PUMA alpha. Suggested positive control: HeLa, NIH-3T3 cell lysate. Immunogen sequence: AAPPLTLEGP VQSHHGTP.
Specifications
Specifications
| Antigen | PUMA delta/gamma |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | BBC3 |
| Gene Accession No. | Q99ML1, Q9BXH1 |
| Gene Alias | BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1 |
| Gene Symbols | BBC3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.