Learn More
Invitrogen™ PUMA alpha Monoclonal Antibody (2A8F6)

Description
A suggested positive control is K562 cell lysate.
Specifications
Specifications
| Antigen | PUMA alpha |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2A8F6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BBC3 |
| Gene Accession No. | Q80ZG6, Q9BXH1 |
| Gene Alias | BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; FLJ42994; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.