Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PUMA delta/gamma Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116382
Description
PUMA delta/gamma Polyclonal Antibody for Western Blot
PUMA is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMA-a and PUMA-b. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis. Antisense inhibition of PUMA expression reduced the apoptotic response to p53, and PUMA is likely to play a role in mediating p53-induced cell death through the cytochrome c/Apaf-1-dependent pathway.
Specifications
PUMA delta/gamma | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9BXH1 | |
BBC3 | |
KLH conjugated synthetic peptide between 158-192 amino acids from human BBC3. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
BBC3 | |
BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
27113 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction