Learn More
Eisco™ Pumice Specimen
SCI_ED
Catalog No. S24766 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24766 1 Ea. 2 to 3 cm
S26597 10/Pk. 2 to 3 cm
S24767 12/Pk. 2 to 3 cm
S26649 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24766 Supplier Eisco™ Supplier No. ESNG0048
Add to Cart
Add to Cart
Pumice is formed quickly during volcanic eruptions around pockets of air within lava.
  • Floats on water due to internal air pockets
  • For use in demonstrating effect of vesicles on rock density
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Glassy texture is result of swift cooling in mid-air
Specifications
Product Type Igneous Rock
For Use With (Application) Demonstrating effect of vesicles on rock density
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Pumice