Description
- Floats on water due to internal air pockets
- For use in demonstrating effect of vesicles on rock density
- Correlates well to Next Generation Science Standards Evidence Statements
- Glassy texture is result of swift cooling in mid-air
Specifications
Specifications
| Product Type | Igneous Rock |
| For Use With (Application) | Demonstrating effect of vesicles on rock density |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Pumice |