Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pumilio 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981178
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1178 Supplier Bioss Supplier No. BS11218R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PUM1 (pumilio homolog 1) is a sequence-specific RNA-binding protein that acts as a post-transcriptional repressor by binding the 3′-UTR of mRNA targets. It is a member of the PUF family comprised of evolutionarily conserved RNA-binding proteins. PUM1 binds to an RNA consensus sequence, the Pumilio Response Element (PRE), and 5′-UGUANAUA-3′ that is related to the Nanos Response Element (NRE). PUM1 mediates post-transcriptonal repression of transcripts via different mechanisms - acts via direct recruitment of the CCR4-POP2-NOT deadenylase leading to translational inhibition and mRNA degradation. Mutations to the gene can result in spinocerebellar ataxia 47.

Specifications

Antigen Pumilio 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PUM1
Gene Accession No. Q14671
Gene Alias AA517475; HsPUM; Kiaa0099; mKIAA0099; PUM1; PUMH; PUMH1; pumilio 1; Pumilio homolog 1; pumilio RNA binding family member 1; pumilio RNA-binding family member 1; pumilio-1; PUML1; Pumm
Gene Symbols PUM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Pumilio 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9698
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.