Learn More
Pumilio 2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300202A
Description
The recommended shelf life for this product is 1 year from date of receipt.
PUM2 is a member of the PUF family, evolutionarily conserved RNA-binding proteins related to the Pumilio proteins of Drosophila and the fem-3 mRNA binding factor proteins of C. elegans. This protein contains a sequence-specific RNA binding domain comprised of eight repeats and N- and C-terminal flanking regions, and serves as a translational regulator of specific mRNAs by binding to their 3′ untranslated regions. The evolutionarily conserved function of this protein in invertebrates and lower vertebrates suggests that the human protein may be involved in translational regulation of embryogenesis, and cell development and differentiation.Specifications
| Pumilio 2 | |
| Polyclonal | |
| Unconjugated | |
| PUM2 | |
| PUM2; KIAA0235; PUMH2; Pumilio homolog 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23369, 80913 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q80U58, Q8TB72 | |
| Pum2 | |
| Between 100 and 150 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.