Learn More
PUNC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA553291
Description
Immunogen sequence: SAELAFAVEP SDDVAVPGQP IVLDCRVEGT PPVRITWRKN GVELPESTHS TLLANGSLMI RHFRLEPGGS PSDEGDYECV AQNRFGLVVS RKARIQAATM SDFHVHPQAT VGEEGGVARF QCQIHGLPKP LITWEKN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.
Specifications
| PUNC | |
| Polyclonal | |
| Unconjugated | |
| IGDCC3 | |
| IGDCC3 | |
| Recombit protein corresponding to Human PUNC | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVU1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9543 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only