Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Punt Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$268.00 - $706.50

Specifications

Antigen Punt
Concentration 0.75 mg/mL
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018166
SDP
View Documents
Novus Biologicals
NB3006620.025MG
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NB300662005
SDP
View Documents
Novus Biologicals
NB3006620.05MG
0.05 mg
Each for $426.00
Only null left
Add to Cart
 
NB300662
SDP
View Documents
Novus Biologicals
NB300662
0.1 mg
Each for $706.50
Only null left
Add to Cart
 
Description

Description

Punt Polyclonal specifically detects Punt in Drosophila samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

Punt
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500
Polyclonal
Rabbit
Activin A receptor 88CD, Activin receptor, Activin receptor type II, ATR, Atr II, Atr88CD, AtrII, CG7904, dlhC, Dorsal holes C, j5A5, l(3)10460, Pun, Punt receptor serine/threonine kinase, Put, STK C, STKC, TGF B, TGF beta, TGF beta l, TGF beta type receptor like, Tgf r, TGFB, TGFbeta
IgG
Punt
0.75 mg/mL
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Drosophila
Synthetic peptide: KSTTEATTQVPKEKTQDGSN[C]. Peptides was amidated for conjugation.
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.