Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Punt Rabbit anti-Drosophila, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc PUNT112AP

Description
Punt Polyclonal antibody specifically detects Punt in Drosophila samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
Punt | |
Polyclonal | |
Unconjugated | |
Act-r; Atr; Atr88CD; AtrII; Atr-II; CG7904; dlhC; Dmel\CG7904; l(3)10460; l(3)j5A5; pun; PUNT; Put; STK-C; TGF-B; TGF-beta; Tgf-r | |
Synthetic peptide corresponding to C terminal amino acids 497-515 of Drosophila melanogaster PUNT protein. Peptides were amidated for conjugation.Sequence:QLLIKNHTNIDDAKESTNC | |
200 μL | |
Primary | |
-20°C | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Drosophila | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction