Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Punt Rabbit anti-Drosophila, Polyclonal, FabGennix

Catalog No. 502208801
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8801 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8801 Supplier Fabgennix Inc Supplier No. PUNT101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Punt Polyclonal antibody specifically detects Punt in Drosophila samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Punt
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Alias Act-r; Atr; Atr88CD; AtrII; Atr-II; CG7904; dlhC; Dmel\CG7904; l(3)10460; l(3)j5A5; pun; PUNT; Put; STK-C; TGF-B; TGF-beta; Tgf-r
Host Species Rabbit
Immunogen N-epitope synthetic peptide corresponding to unique amino acid sequences on D. melanogaster Punt protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Drosophila
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.