Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PURA Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP192651IR
Description
PURA Polyclonal antibody specifically detects PURA in Human, Mouse samples. It is validated for Western BlotSpecifications
PURA | |
Polyclonal | |
Western Blot | |
PUR1PUR-ALPHA, PURALPHA, purine-rich element binding protein A, Purine-rich single-stranded DNA-binding protein alpha, transcriptional activator protein Pur-alpha | |
The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Purine-Rich Element Binding Protein Alpha using the numbering given in entry NP_005850.1 (GeneID 5813). | |
0.1 mL | |
Cell Cycle and Replication | |
5813 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction