Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PURA Monoclonal Antibody (1D6)
GREENER_CHOICE

Catalog No. PIMA519766
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519766 Supplier Invitrogen™ Supplier No. MA519766
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: TQGQTIALPA QGLIEFRDAL AKLIDDYGVE EEPAELPEGT SLTVDNKRFF FDVGSNKYGV FMRVSEVKPT YRNSITVPYK VWAKFGHTFC KYSEETKKIQ EKQREKRAAC.

This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PURA
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PURA
Gene Accession No. Q00577
Gene Alias CAGER-1; MRD31; Pur alpha; PUR1; Pura; pura {ECO:0000250; PURALPHA; Pur-alpha; purine rich element binding protein A; purine-rich element binding protein A; purine-rich single-stranded DNA-binding protein alpha; purine-rich single-stranded DNA-binding protein alpha {ECO:0000250; ssCRE-BP; transcriptional activator protein Pur-alpha; transcriptional activator protein Pur-alpha {ECO:0000250; UniProtKB:P42669}
Gene Symbols PURA
Host Species Mouse
Immunogen PURA (NP_005850, 183 a.a. ~ 292 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5813
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.