Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PURA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115808 Supplier Invitrogen™ Supplier No. PA5115808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PURA.

This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PURA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PURA
Gene Accession No. P42669, P86252, Q00577
Gene Alias CAGER-1; MRD31; Pur alpha; PUR1; Pura; pura {ECO:0000250; PURALPHA; Pur-alpha; purine rich element binding protein A; purine-rich element binding protein A; purine-rich single-stranded DNA-binding protein alpha; purine-rich single-stranded DNA-binding protein alpha {ECO:0000250; ssCRE-BP; transcriptional activator protein Pur-alpha; transcriptional activator protein Pur-alpha {ECO:0000250; UniProtKB:P42669}
Gene Symbols PURA
Host Species Rabbit
Immunogen A synthesized peptide derived from human PURA(Accession Q00577), corresponding to amino acid residues V106-Y156.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19290, 307498, 5813
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.