Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puratrophin 1/PLEKHG4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981634
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1634 Supplier Bioss Supplier No. BS11747R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Possible role in intracellular signaling and cytoskeleton dynamics at the Golgi.

Specifications

Antigen Puratrophin 1/PLEKHG4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLEKHG4
Gene Accession No. Q58EX7
Gene Alias ARHGEF44; PH domain-containing family G member 4; pleckstrin homology and RhoGEF domain containing G4; pleckstrin homology domain containing, family G (with RhoGef domain) member 4; Pleckstrin homology domain-containing family G member 4; PLEKHG4; PRTPHN1; Puratrophin-1; Purkinje cell atrophy associated protein 1; Purkinje cell atrophy-associated protein 1; SCA4
Gene Symbols PLEKHG4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Puratrophin 1/PLEKHG4 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25894
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.