Learn More
BUCHI Pure Essential Chromatography System
SDP
Catalog No. 05405930 Shop All BUCHI Products
Click to view available options
Style:
Pump: No; Detector: No; Fraction Collector: Yes
Pump: No; Detector: Yes; Fraction Collector: No
Pump: Yes; Detector: No; Fraction Collector: No
Pump: Yes; Detector: Yes; Fraction Collector: No
Pump: Yes; Detector: Yes; Fraction Collector: Yes
This system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
  • Benefits from a basic design and setup
  • The small footprint of the instrument allows for placement even in crowded labs, and its robustness makes it highly reliable
  • Basic but powerful and safe thanks to a 50 bar (725 psi) pump, pressure sensors, and regulators
  • Expansion possibilities include the addition of a UV detector and fraction collector to the Pure Chromatography C-900
  • There are no limits regarding the size of the collected fractions
  • Additional options allow you to be flexible regarding sample loading (liquid and solid) and the cartridge holder (any size between 4 – 5000 g)
  • Intuitive navigation provided by novel and state-of-the-art software (methods, run data, and general settings)
  • All tubing and parts of the instrument are easily accessible and allow the user to follow and monitor the process flow
  • Automation simplifies the entire separation process
Specifications
Style Pump: No; Detector: No; Fraction Collector: Yes
Type Flash Chromatography Kit I
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.